1-(Triphenylmethyl)-L-histidine
Supplier: BACHEM AMERICAS INC
Synonyms:
H-His(Trt)-OH, 1-Trityl-L-histidine, H-His(1-Trt)-OH
4011443.0005
4011443.0025
E-2980.0005BAEA
284.04
CAD
E-2980.0005BA
E-2980.0025BA
1-(Triphenylmethyl)-L-histidine
1-(Triphenylmethyl)-L-histidine
Sequence: H-His(1-Trt)-OH
The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.
Synonym(s): H-His(τ-Trt)-OH
Formula:
C₂₅H₂₃N₃O₂ MW: 397.478 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00153444 CAS Number: 35146-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...