H-Trp(Boc)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-2-Amino-3-(1-(tert-butoxycarbonyl)-1H-indol-3-yl)propanoic acid

4017673.0005 4017673.0025
E-2975.0005BAEA 490.51 CAD
E-2975.0005BA E-2975.0025BA
H-Trp(Boc)-OH
H-Trp(Boc)-OH

Sequence: H-Trp(Boc)-OH

Formula: C₁₆H₂₀N₂O₄
MW: 304.346 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237294
CAS Number: 146645-63-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR