(Ac-Cys-OH)₂

Supplier: BACHEM AMERICAS INC

Synonyms: N,N'-Diacetyl-L-Cystine

4017817.0001 4017817.0005
E-2965.0001BAEA 605.93 CAD
E-2965.0001BA E-2965.0005BA
(Ac-Cys-OH)₂
(Ac-Cys-OH)₂

Sequence: (Ac-Cys-OH)₂


As the disulfide bond can be cleaved under mild conditions (e.g. with trialkylphosphines) cystine derivatives are highly economic educts for synthesizing Cys derivatives. The cystine derivatives of fluorophores (e.g. J-1175) or the pNA derivatives may be chosen as educts for synthesizing Cys substrates.


Synonym(s): DiNAC

Formula: C₁₀H₁₆N₂O₆S₂
Storage Temperature: Freezer
MDL Number: MFCD00152053
CAS Number: 5545-17-5
EINECS: 226-911-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR