L-Tyrosine benzyl ester

Supplier: BACHEM AMERICAS INC

4017661.0005 4017661.0025 4017661.0100
E-2955.0005BAEA 180.91 CAD
E-2955.0005BA E-2955.0025BA E-2955.0100BA
L-Tyrosine benzyl ester

Sequence: H-Tyr-OBzl


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₆H₁₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD00191034
CAS Number: 42406-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR