(S)-Benzyl-2-amino-3-hydroxypropanoate
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Ser-OBzl
4004981.0005
4004981.0025
4004981.0100
E-2950.0005BAEA
189.95
CAD
E-2950.0005BA
E-2950.0025BA
E-2950.0100BA
(S)-Benzyl-2-amino-3-hydroxypropanoate
(S)-Benzyl-2-amino-3-hydroxypropanoate
Sequence: H-Ser-OBzl
Bachem also offers derivatives of phosphoserine and sulfoserine.
Formula:
C₁₀H₁₃NO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD00038955 CAS Number: 1738-72-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...