(R)-2-Amino-3-(tritylthio)propanamide

Supplier: BACHEM AMERICAS INC

Synonyms: H-Cys(Trt)-NH2, H-Cys(Trt)-NH₂, S-Trityl-L-cysteinamide

4015634.0001 4015634.0005
E-2930.0001BAEA 180.91 CAD
E-2930.0001BA E-2930.0005BA
(R)-2-Amino-3-(tritylthio)propanamide
(R)-2-Amino-3-(tritylthio)propanamide

Sequence: H-Cys(Trt)-NH₂

Formula: C₂₂H₂₂N₂OS
MW: 362.49 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22126061
CAS Number: 166737-85-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR