Abz-Gly-OH·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: 2-(2-Aminobenzamido)acetic acid hydrochlorideAbz-Gly-OH·HCl

4015644.0005 4015644.0025
E-2920.0025BAEA 7226.04 CAD
E-2920.0025BA E-2920.0005BA
Abz-Gly-OH·HCl
Abz-Gly-OH·HCl

Sequence: Abz-Gly-OH · HCl


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.


Synonym(s): 2-Aminohippuric acid · HCl

Formula: C₉H₁₁ClN₂O₃
MW: 230.648 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00237464
CAS Number: 256657-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR