N-Tosyl-l-proline

Supplier: BACHEM AMERICAS INC

Synonyms: Tos-Pro-OH, (2S)-1-[(4-methylphenyl)sulphonyl]pyrrolidine-2-carboxylic acid, N-Toluenesulfonyl-L-proline, N-Toluenesulphonyl-L-proline, (2S)-1-[(4-methylphenyl)sulfonyl]pyrrolidine-2-carboxylic acid

4017504.0025 4017504.0100
E-2915.0025BAEA 361.41 CAD
E-2915.0025BA E-2915.0100BA
N-Tosyl-l-proline
N-Tosyl-l-proline

Sequence: Tos-Pro-OH

Formula: C₁₂H₁₅NO₄S
MW: 269.315 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00238392
CAS Number: 51077-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR