H-Arg(Pmc)-OH

Supplier: BACHEM AMERICAS INC

4013348.0001 4013348.0005 4013348.0025
E-2885.0001BAEA 180.91 CAD
E-2885.0001BA E-2885.0005BA E-2885.0025BA
H-Arg(Pmc)-OH
H-Arg(Pmc)-OH

Sequence: H-Arg(Pmc)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.
See also F-1870, H-Arg(NO₂)-OH, in the subfamily 'Arginine Substrates and Inhibitors'.

Formula: C₂₀H₃₂N₄O₅S
Storage Temperature: Ambient
MDL Number: MFCD00153419
CAS Number: 112160-37-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR