1-Methyl-L-glutamate

Supplier: BACHEM AMERICAS INC

Synonyms: H-Glu-OMe, 1-Methyl L-Glutamate, L-Glutamic Acid 1-Methyl Ester

4011104.0001 4011104.0005
E-2875.0001BAEA 206.51 CAD
E-2875.0001BA E-2875.0005BA
1-Methyl-L-glutamate
1-Methyl-L-glutamate

Sequence: H-Glu-OMe


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₆H₁₁NO₄
MW: 161.16 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00171677
CAS Number: 6384-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR