H-His(3-Me)-OH

Supplier: BACHEM AMERICAS INC

4000900.0250 4000900.1000
E-2845.0250BAEA 211.67 CAD
E-2845.0250BA E-2845.1000BA
H-His(3-Me)-OH
H-His(3-Me)-OH

Sequence: H-His(3-Me)-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Synonym(s): H-His(π-Me)-OH

Formula: C₇H₁₁N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00083658
CAS Number: 368-16-1
EINECS: 206-704-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR