N-(Triphenylmethyl)glycine

Supplier: BACHEM AMERICAS INC

Synonyms: 2-(Tritylamino)acetic acid, Trt-Gly-OH, N-Tritylglycine

4014956.0005 4014956.0025 4014956.0100
E-2835.0005BAEA 180.91 CAD
E-2835.0005BA E-2835.0025BA E-2835.0100BA
N-(Triphenylmethyl)glycine
N-(Triphenylmethyl)glycine

Sequence: Trt-Gly-OH


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: C₂₁H₁₉NO₂
Storage Temperature: Freezer
MDL Number: MFCD00004276
CAS Number: 5893-05-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR