H-Asn(Trt)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nγ-Trityl-L-Asparagine, Ngamma-Trityl-L-asparagine, (S)-2-Amino-4-oxo-4-(tritylamino)butyric acid

4014946.0001 4014946.0005
E-2810.0001BAEA 180.91 CAD
E-2810.0001BA E-2810.0005BA
H-Asn(Trt)-OH
H-Asn(Trt)-OH

Sequence: H-Asn(Trt)-OH


For isoasparagine derivatives please see the subfamily "Isoasparagine" filed under "Aspartic Acid".

Formula: C₂₃H₂₂N₂O₃
MW: 374.430 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153422
CAS Number: 132388-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR