L-Glutamic acid 5-ethyl ester

Supplier: BACHEM AMERICAS INC

Synonyms: 5-Ethyl L-Glutamate, L-Glutamic acid 5-Ethyl ester, H-Glu(OEt)-OH

4014122.0025 4014122.0100
E-2780.0025BAEA 439.41 CAD
E-2780.0025BA E-2780.0100BA
L-Glutamic acid 5-ethyl ester
L-Glutamic acid 5-ethyl ester

Sequence: H-Glu(OEt)-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₇H₁₃NO₄
MW: 175.2 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037217
CAS Number: 1119-33-1
EINECS: 214-274-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR