H-Cys(BZL)-OET·HCl
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Cys(Bzl)-OEt·HCl
4012395.0005
4012395.0025
4012395.0100
E-2720.0005BAEA
180.91
CAD
E-2720.0005BA
E-2720.0025BA
E-2720.0100BA
H-Cys(BZL)-OET·HCl
H-Cys(BZL)-OET·HCl
Sequence: H-Cys(Bzl)-OEt · HCl
Formula:
C₁₂H₁₈ClNO₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD00038909 CAS Number: 52844-67-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...