H-Cys(StBu)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: S-tert-Butylmercapto-L-cysteine

4030538.0005 4030538.0025
E-2710.0005BAEA 180.91 CAD
E-2710.0005BA E-2710.0025BA
H-Cys(StBu)-OH
H-Cys(StBu)-OH

Sequence: H-Cys(StBu)-OH

Formula: C₇H₁₅NO₂S₂
MW: 209.322 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00002609
CAS Number: 30044-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR