4-Hydroxyhippuric acid

Supplier: BACHEM AMERICAS INC

Synonyms: 2-(4-Hydroxybenzamido)acetic acid4-Hydroxy-hippuric acid

4005059.0001 4005059.0005
E-2655.0005BAEA 1140.87 CAD
E-2655.0005BA E-2655.0001BA
4-Hydroxyhippuric acid
4-Hydroxyhippuric acid

Sequence: 4-Hydroxy-hippuric acid


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.


Synonym(s): 4-Hydroxy-Bz-Gly-OH

Formula: C₉H₉NO₄
MW: 195.174 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237117
CAS Number: 2482-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR