Leucinamide

Supplier: BACHEM AMERICAS INC

Synonyms: H-Leu-NH₂

4019083.0005 4019083.0025
E-2650.0005BAEA 180.91 CAD
E-2650.0005BA E-2650.0025BA
Leucinamide
Leucinamide

Sequence: H-Leu-NH₂


Bachem additionally offers deuterated and ¹⁵N-labeled leucine: Fmoc-[D₁₀]Leu-OH (B-4120) and Fmoc-[¹⁵N]Leu-OH (B-2655).


Substrate for leucine aminopeptidase.

Formula: C₆H₁₄N₂O
Storage Temperature: Freezer
MDL Number: MFCD00238027
CAS Number: 687-51-4
EINECS: 211-696-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR