Ethyl L-histidinate·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-His-OEt·2 HCl

4007084.0025 4007084.0100
E-2630.0025BAEA 317.74 CAD
E-2630.0025BA E-2630.0100BA
Ethyl L-histidinate·HCl
Ethyl L-histidinate·HCl

Sequence: H-His-OEt · 2 HCl


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₈H₁₅Cl₂N₃O₂
MW: 256.127 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00050627
CAS Number: 93923-84-3
EINECS: 300-174-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR