Nε-trifluoroacetyl-L-lysine
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Lys(Tfa)-OH, ε-TFA-lysine, (S)-2-Amino-6-(2,2,2-trifluoroacetamido)hexanoic acid
4008182.0005
4008182.0025
4008182.0100
E-2615.0005BAEA
185.46
CAD
E-2615.0005BA
E-2615.0025BA
E-2615.0100BA
Nε-trifluoroacetyl-L-lysine
Nepsilon-trifluoroacetyl-L-lysine
Sequence: H-Lys(Tfa)-OH
For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".
Formula:
C₈H₁₃F₃N₂O₃ MW: 242.20 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00037223 CAS Number: 10009-20-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...