L-Valine methyl ester hydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Val-OMe.HCl, L-2-Aminoisovaleric Acid Methyl Ester Hydrochloride, L-Methyl valinate hydrochloride, H-Val-OMe·HCl
4002561.0025
4002561.0100
E-2595.0025BAEA
180.91
CAD
E-2595.0025BA
E-2595.0100BA
L-Valine methyl ester hydrochloride
L-Valine methyl ester hydrochloride
Sequence: H-Val-OMe · HCl
Bachem additionally offers Fmoc-[¹⁵N]Val-OH, B-2670.
Formula:
C₆H₁₄ClNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00012497 CAS Number: 6306-52-1 EINECS: 228-620-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...