L-Tyrosine ethyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Tyr-OEt·HCl, H-Tyr-OEt.HCl

4000537.0025 4000537.0100
E-2550.0025BAEA 180.91 CAD
E-2550.0025BA E-2550.0100BA
L-Tyrosine ethyl ester hydrochloride
L-Tyrosine ethyl ester hydrochloride

Sequence: H-Tyr-OEt · HCl


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₁H₁₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00063047
CAS Number: 4089-07-0
EINECS: 223-820-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR