L-Tyrosine-tert-butyl ester

Supplier: BACHEM AMERICAS INC

4001023.0005 4001023.0025
E-2535.0005BAEA 206.51 CAD
E-2535.0005BA E-2535.0025BA
L-Tyrosine-tert-butyl ester
L-Tyrosine-tert-butyl ester

Sequence: H-Tyr-OtBu


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.

Formula: C₁₃H₁₉NO₃
Storage Temperature: Freezer
MDL Number: MFCD00042644
CAS Number: 16874-12-7
EINECS: 240-902-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR