L(-)-Tyrosine
Supplier: BACHEM AMERICAS INC
Synonyms:
L-Tyrosine, (S)-(-)-Tyrosine, Tyrosine, H-Tyr-OH
4031323.0250
4031323.1000
E-2515.0250BAEA
250.43
CAD
E-2515.0250BA
E-2515.1000BA
L(-)-Tyrosine
L(-)-Tyrosine
Sequence: H-Tyr-OH
Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.
Formula:
C₉H₁₁NO₃ MW: 181.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002606 CAS Number: 60-18-4 EINECS: 200-460-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...