L-Tryptophan ethyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Trp-OEt.HCl, H-Trp-OEt·HCl, Ethyl L-tryptophanate hydrochloride, (S)-2-Amino-3-(3-indolyl)propionic Acid Ethyl Ester Hydrochloride

4002156.0025 4002156.0100
E-2510.0025BAEA 206.51 CAD
E-2510.0025BA E-2510.0100BA
L-Tryptophan ethyl ester hydrochloride
L-Tryptophan ethyl ester hydrochloride

Sequence: H-Trp-OEt · HCl

Formula: C₁₃H₁₆N₂O₂
MW: 268.74 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038993
CAS Number: 2899-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR