L(-)-Tryptophan
Supplier: BACHEM AMERICAS INC
Synonyms:
L-Tryptophan, Tryptophan, L-α-Amino-3-indolepropionic acid, (S)-2-Amino-3-(3-indolyl)propionic acid, H-Trp-OH
4031215.0100
4031215.0250
4031215.1000
E-2500.0100BAEA
175.66
CAD
E-2500.0100BA
E-2500.0250BA
E-2500.1000BA
L(-)-Tryptophan
L(-)-Tryptophan
Sequence: H-Trp-OH
Formula:
C₁₁H₁₂N₂O₂ MW: 204.23 g/mol Melting Pt: 290 Storage Temperature: Ambient |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 Merck Index: 12,09929 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...