S-Trityl-L-cysteine
Supplier: BACHEM AMERICAS INC
Synonyms:
H-Cys(Trt)-OH
4001485.0025
4001485.0100
E-2495.0025BAEA
258.14
CAD
E-2495.0025BA
E-2495.0100BA
S-Trityl-L-cysteine
S-Trityl-L-cysteine
Sequence: H-Cys(Trt)-OH
Synonym(s): STLC
Cys(Trt)-containing peptides can be converted in high yields into the corresponding cystine peptides by iodine oxidation. Cys(Trt) (STLC, NSC 83265) is a reversible, tight binding inhibitor of the human kinesin Eg5 that specifically blocks mitotic progression.
Formula:
C₂₂H₂₁NO₂S MW: 363.5 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00002611 CAS Number: 2799-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...