Methyl-L-threoninate hydrochloride

Supplier: BACHEM AMERICAS INC

4000503.0005 4000503.0025
E-2455.0005BAEA 185.46 CAD
E-2455.0005BA E-2455.0025BA
Methyl-L-threoninate hydrochloride
Methyl-L-threoninate hydrochloride

Sequence: H-Thr-OMe · HCl


Bachem also offers phosphothreonine derivatives.

Formula: C₅H₁₂ClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD00037677
CAS Number: 39994-75-7
EINECS: 254-738-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR