H-His(Bzl)-OH

Supplier: BACHEM AMERICAS INC

4007055.0005 4007055.0025
E-2435.0005BAEA 284.04 CAD
E-2435.0005BA E-2435.0025BA
H-His(Bzl)-OH
H-His(Bzl)-OH

Mix of τ-Bzl (mainly formed) and π-Bzl isomer. CAS Numbers: 16832-24-9 (τ-isomer), 26086-41-9 (π-isomer).


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.


Mix of τ-Bzl (mainly formed) and π-Bzl isomer. CAS Numbers: 16832-24-9 (τ-isomer), 26086-41-9 (π-isomer).

Formula: C₁₃H₁₅N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD00038630
CAS Number: 16832-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR