L-Threonine benzyl ester oxalate

Supplier: BACHEM AMERICAS INC

4001337.0005 4001337.0025
E-2430.0005BAEA 180.91 CAD
E-2430.0005BA E-2430.0025BA
L-Threonine benzyl ester oxalate
L-Threonine benzyl ester oxalate

Sequence: H-Thr-OBzl · oxalate (1:1)


Bachem also offers phosphothreonine derivatives.

Formula: C₁₃H₁₇NO₇
Storage Temperature: Ambient
MDL Number: MFCD02259509
CAS Number: 201274-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR