L-Threoninamide hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Thr-NH₂·HCl, (2S,3R)-2-Amino-3-hydroxybutanamide hydrochloride

4004473.0001 4004473.0005 4004473.0025
E-2425.0001BAEA 180.83 CAD
E-2425.0001BA E-2425.0005BA E-2425.0025BA
L-Threoninamide hydrochloride
L-Threoninamide hydrochloride

Sequence: H-Thr-NH₂ · HCl


Bachem also offers phosphothreonine derivatives.

Formula: C₄H₁₁ClN₂O₂
MW: 154.594 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058288
CAS Number: 33209-01-7
EINECS: 251-407-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR