N-(Trifluoroacetyl)glycine

Supplier: BACHEM AMERICAS INC

Synonyms: (2,2,2-Trifluoroacetyl)glycine, Tfa-Gly-OH

4002918.0005 4002918.0025
E-2415.0005BAEA 439.41 CAD
E-2415.0005BA E-2415.0025BA
N-(Trifluoroacetyl)glycine
N-(Trifluoroacetyl)glycine

Sequence: Tfa-Gly-OH


Bachem additionally offers deuterated glycine: Fmoc-[D₂]Gly-OH B-4125. For N-methylglycine derivatives see Sarcosine.

Formula: C₄H₄F₃NO₃
MW: 171.075 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00029784
CAS Number: 383-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR