L(+)-Serine

Supplier: BACHEM AMERICAS INC

Synonyms: H-Ser-OH, (S)-2-Amino-3-hydroxypropanoic acid, Serine, (S)-(+)-Serine

4031220.0100 4031220.0250 4031220.1000
E-2375.0100BAEA 175.66 CAD
E-2375.0100BA E-2375.0250BA E-2375.1000BA
L(+)-Serine
L(+)-Serine

Sequence: H-Ser-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 259
Storage Temperature: Ambient
MDL Number: MFCD00064224
CAS Number: 56-45-1
EINECS: 200-274-3
Merck Index: 14,00846

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR