N,N-dimethyl-2-(methylamino)acetamide

Supplier: BACHEM AMERICAS INC

4001134.0001 4001134.0005
E-2365.0001BAEA 291.08 CAD
E-2365.0001BA E-2365.0005BA
N,N-dimethyl-2-(methylamino)acetamide
N,N-dimethyl-2-(methylamino)acetamide

Sequence: Sar-NMe₂


Bachem also offers Fmoc-Sar-Wang resin (D-1805) and Fmoc-Sar-SASRIN resin (-D-1645).


Synonym(s): N-Me-Gly-NMe₂

Formula: C₅H₁₂N₂O
Storage Temperature: Freezer
MDL Number: MFCD00080937
CAS Number: 1857-20-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR