L-Pyroglutamic acid pentachlorophenyl ester

Supplier: BACHEM AMERICAS INC

Synonyms: Pyr-OPcp

4000759.0001 4000759.0005
E-2350.0001BAEA 180.91 CAD
E-2350.0001BA E-2350.0005BA
L-Pyroglutamic acid pentachlorophenyl ester
L-Pyroglutamic acid pentachlorophenyl ester

Sequence: Pyr-OPcp


Also called 5-oxoproline or pidolic acid. Bachem uses the acronym Pyr for denoting pyroglutamic acid. Further abbreviations as Glp, Pgl, pGlu (pE), or < Glu (< E) are also in use.

Formula: C₁₁H₆Cl₅NO₃
MW: 377.423 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00057185
CAS Number: 28990-85-4
EINECS: 249-360-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR