L-Proline methyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Pro-OMe.HCl, H-Pro-OMe·HCl

4000661.0025 4000661.0100
E-2325.0025BAEA 317.74 CAD
E-2325.0025BA E-2325.0100BA
L-Proline methyl ester hydrochloride
L-Proline methyl ester hydrochloride

Sequence: H-Pro-OMe · HCl

Formula: C₆H₁₁NO₂·HCl
MW: 165.6 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00012708
CAS Number: 2133-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR