L-Proline-tert-butyl ester

Supplier: BACHEM AMERICAS INC

4002617.0005 4002617.0025
E-2310.0005BAEA 206.51 CAD
E-2310.0005BA E-2310.0025BA
L-Proline-tert-butyl ester
L-Proline-tert-butyl ester

Sequence: H-Pro-OtBu (syrup)

Formula: C₉H₁₇NO₂
Storage Temperature: Freezer
MDL Number: MFCD00037879
CAS Number: 2812-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR