L-Proline benzyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Pro-OBzl.HCl

4002624.0005 4002624.0025
E-2305.0005BAEA 180.91 CAD
E-2305.0005BA E-2305.0025BA
L-Proline benzyl ester hydrochloride
L-Proline benzyl ester hydrochloride

Sequence: H-Pro-OBzl · HCl

Formula: C₁₂H₁₅NO₂
MW: 241.72 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00039002
CAS Number: 16652-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR