L-Prolinamide hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: (S)-Pyrrolidine-2-carboxamide hydrochloride, H-Pro-NH₂·HCl

4000855.0005 4000855.0025
E-2300.0005BAEA 206.51 CAD
E-2300.0005BA E-2300.0025BA
L-Prolinamide hydrochloride
L-Prolinamide hydrochloride

Sequence: H-Pro-NH₂ · HCl

Formula: C₅H₁₁ClN₂O
Storage Temperature: Freezer
MDL Number: MFCD00058156
CAS Number: 42429-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR