L(-)-Proline

Supplier: BACHEM AMERICAS INC

4031088.0100 4031088.0500
E-2290.0100BAEA 180.14 CAD
E-2290.0100BA E-2290.0500BA
L(-)-Proline
L(-)-Proline

Sequence: H-Pro-OH

Formula: C₅H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD00064318
CAS Number: 147-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR