(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride
Supplier: BACHEM AMERICAS INC
Synonyms:
L-Methyl phenylalanate hydrochloride, L-Phenylalanine methyl ester hydrochloride, H-Phe-OMe·HCl
4000658.0025
4000658.0100
E-2270.0025BAEA
180.91
CAD
E-2270.0025BA
E-2270.0100BA
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride
(S)-Methyl-2-amino-3-phenylpropanoate hydrochloride
Sequence: H-Phe-OMe · HCl
Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Formula:
C₁₀H₁₄ClNO₂ MW: 215.68 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00012489 CAS Number: 7524-50-7 EINECS: 231-383-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...