L-Phenylalanine-tert-butyl ester hydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Phe-OtBu·HCl, H-Phe-OtBu.HCl

4001056.0005 4001056.0025
E-2255.0005BAEA 211.61 CAD
E-2255.0005BA E-2255.0025BA
L-Phenylalanine-tert-butyl ester hydrochloride
L-Phenylalanine-tert-butyl ester hydrochloride

Sequence: H-Phe-OtBu · HCl


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.


CAS number (net): 16874-17-2.

Formula: C₁₃H₂₀ClNO₂
MW: 257.758 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038894
CAS Number: 15100-75-1
EINECS: 239-151-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR