N-Methyl-L-serine

Supplier: BACHEM AMERICAS INC

4002644.0001 4002644.0005
E-2190.0001BAEA 903.62 CAD
E-2190.0001BA E-2190.0005BA
N-Methyl-L-serine
N-Methyl-L-serine

Sequence: N-Me-Ser-OH


Bachem also offers derivatives of phosphoserine and sulfoserine.

Formula: C₄H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD00037756
CAS Number: 2480-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR