N-Me-Phe-OMe·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: N-Me-Phe-OMe·HCl

4002105.0001 4002105.0005
E-2170.0001BAEA 593.81 CAD
E-2170.0001BA E-2170.0005BA
N-Me-Phe-OMe·HCl
N-Me-Phe-OMe·HCl

Sequence: N-Me-Phe-OMe · HCl


Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Short N-methyl-phenylalanine-rich peptides were shown to function as versatile blood-brain barrier shuttles.

Formula: C₁₁H₁₆ClNO₂
MW: 229.704 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083661
CAS Number: 19460-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR