N-Methylphenylalanine
Supplier: BACHEM AMERICAS INC
Synonyms:
N-Me-Phe-OH
4000572.0001
4000572.0005
E-2165.0001BAEA
183.09
CAD
E-2165.0001BA
E-2165.0005BA
N-Methylphenylalanine
N-Methylphenylalanine
Sequence: N-Me-Phe-OH
Bachem additionally offers deuterated phenylalanine: Fmoc-[ring-D₅]Phe-OH B-4115.
Short N-methyl-phenylalanine-rich peptides were shown to function as versatile blood-brain barrier shuttles.
Formula:
C₁₀H₁₃NO₂ MW: 179.219 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037757 CAS Number: 2566-30-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...