N,O-Dimethyltyrosine

Supplier: BACHEM AMERICAS INC

Synonyms: N-Me-Tyr(Me)-OH

4008161.0250 4008161.1000
E-2160.0250BAEA 232.35 CAD
E-2160.0250BA E-2160.1000BA
N,O-Dimethyltyrosine
N,O-Dimethyltyrosine

Sequence: N-Me-Tyr(Me)-OH


Besides a large selection of tyrosine derivatives, Bachem offers derivatives of O-alkylated tyrosines, thyronine, phosphotyrosine and sulfotyrosine. Bachem additionally offers H-[¹⁵N]Tyr-OH, E-3245.


Synonym(s): N-Me-4-methoxy-Phe-OH

Formula: C₁₁H₁₅NO₃
MW: 209.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00038797
CAS Number: 52939-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR