H-Lys(Me)-OH·HCl

Supplier: BACHEM AMERICAS INC

Synonyms: H-Lys(Me)-OH·HCl

4003843.0250 4003843.1000 4003843.5000
E-2155.0250BAEA 185.3 CAD
E-2155.0250BA E-2155.1000BA E-2155.5000BA
H-Lys(Me)-OH·HCl
H-Lys(Me)-OH·HCl

Sequence: H-Lys(Me)-OH · HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₇H₁₇ClN₂O₂
MW: 196.675 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012621
CAS Number: 7622-29-9
EINECS: 231-539-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR