N-Me-Glu-OH

Supplier: BACHEM AMERICAS INC

Synonyms: N-Methyl-L-glutamic acid

4002660.0001 4002660.0005
E-2130.0001BAEA 258.14 CAD
E-2130.0001BA E-2130.0005BA
N-Me-Glu-OH
N-Me-Glu-OH

Sequence: N-Me-Glu-OH


The derivatives of Glu-NH₂ are assembled in the "Isoglutamine" subfamily.

Formula: C₆H₁₁NO₄
MW: 161.157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065920
CAS Number: 6753-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR