H-Cys(pMeOBzl)-OH
Supplier: BACHEM AMERICAS INC
Synonyms:
S-(4-Methoxybenzyl)-L-cysteine, H-Cys(Mob)-OH
4000523.0025
4000523.0100
E-2120.0025BAEA
1158.05
CAD
E-2120.0025BA
E-2120.0100BA
H-Cys(pMeOBzl)-OH
H-Cys(pMeOBzl)-OH
Sequence: H-Cys(Mob)-OH
Formula:
C₁₁H₁₅NO₃S MW: 241.305 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00037207 CAS Number: 2544-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...