L-Lysine methyl ester dihydrochloride

Supplier: BACHEM AMERICAS INC

Synonyms: H-Lys-OMe·2HCl, H-Lys-OMe.2HCl

4002480.0050 4002480.0250
E-2095.0050BAEA 258.14 CAD
E-2095.0050BA E-2095.0250BA
L-Lysine methyl ester dihydrochloride
L-Lysine methyl ester dihydrochloride

Sequence: H-Lys-OMe · 2 HCl


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₇H₁₆N₂O₂
MW: 233.13 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00039067
CAS Number: 26348-70-9
EINECS: 247-625-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR